Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB169610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169610 100 μg
NB169611 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. NB169610 Supplier Novus Biologicals Supplier No. NBP317267100UL
Add to Cart
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Rabbit Polyclonal Antibody

KCNK9 Polyclonal antibody specifically detects KCNK9 in Human samples. It is validated for Immunofluorescence

Specifications

Antigen KCNK9
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias KT3.2, potassium channel, subfamily K, member 9, TASK-3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: LESDHEMREEEKLKAEEIRIKGKYNISSEDYRQLELVILQSEP
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 51305
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.