Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNMB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB093520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093520 100 μL
NB093521 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093520 Supplier Novus Biologicals Supplier No. NBP305145100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNMB2 Polyclonal specifically detects KCNMB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen KCNMB2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BK channel subunit beta-2, BKbeta2, calcium-activated potassium channel subunit beta-2, Calcium-activated potassium channel, subfamily M subunit beta-2, Charybdotoxin receptor subunit beta-2, hbeta2, Hbeta3, k(VCA)beta-2, large conductance calcium-activated potassium channel beta 2 subunit, large-conductance Ca2+-activated K+ channel beta2 subunit, Maxi K channel subunit beta-2, MaxiK channel beta 2 subunit, MGC22431, potassium large conductance calcium-activated channel, subfamily M, beta member2, slo-beta-2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 68-194 of human KCNMB2 (NP_005823.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10242
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.