Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNMB3 Antibody (S40B-18), Novus Biologicals™
SDP

Catalog No. NBP212916 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212916 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212916 Supplier Novus Biologicals Supplier No. NBP212916

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody has been used in 2 publications

KCNMB3 Monoclonal specifically detects KCNMB3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.

Specifications

Antigen KCNMB3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S40B-18
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 0.1-1.0 ug/ml
Gene Alias BK channel subunit beta-3, BKbeta3, HBETA3, KCNMB2, KCNMBL, potassium large conductance calcium-activated channel, subfamily M beta member3, slo-beta-3, subfamily M subunit beta-3, voltage and Ca2+ activated potassium channel Maxi K beta 3subunit
Gene Symbols KCNMB3
Host Species Mouse
Immunogen Fusion protein amino acids 1-49 (unique N-terminus) of mouse BKBeta3a
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 27094
Test Specificity Detects approx 32 kDa. No cross-reactivity against BKBeta1, BKBeta2, BKBeta3b or BKBeta4.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.