Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNMB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567237 Supplier Invitrogen™ Supplier No. PA567237
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CWLSPALQDLQ ATEANCTVLS VQQIGEVFEC TFTCGADCRG TSQYPCVQV Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which slows activation kinetics, leads to steeper calcium sensitivity, and shifts the voltage range of current activation to more negative potentials than does the beta 1 subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNMB4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNMB4
Gene Accession No. Q86W47
Gene Alias 1700058G18Rik; 2900045G12Rik; big potassium channel beta subunit 4; BK channel beta subunit 4; BK channel subunit beta-4; BKbeta4; calcium activated potassium channel beta 4 subunit; calcium-activated potassium channel subunit beta-4; Calcium-activated potassium channel, subfamily M subunit beta-4; charybdotoxin receptor subunit beta-4; hbeta4; k(VCA)beta-4; KCMB4; KCNMB4; large conductance calcium-dependent potassium ion channel beta 4 subunit; maxi K channel subunit beta-4; MaxiK channel beta-subunit 4; potassium calcium-activated channel subfamily M regulatory beta subunit 4; potassium channel subfamily M regulatory beta subunit 4; potassium large conductance calcium-activated channel, subfamily M, beta member 4; slo-beta-4; Slowpoke beta 4
Gene Symbols KCNMB4
Host Species Rabbit
Immunogen Recombinant Human KCNMB4. Recombinant protein control fragment (Product #RP-107871).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27345
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.