Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNT2 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: S11-33, Novus Biologicals™
SDP

Catalog No. NBP147606FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147606FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147606FR Supplier Novus Biologicals Supplier No. NBP147606FR

Mouse Monoclonal Antibody

KCNT2 Monoclonal specifically detects KCNT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen KCNT2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S11-33
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q6UVM3
Gene Alias KCa4.2, MGC119610, MGC119611, MGC119612, potassium channel subfamily T member 2, potassium channel, subfamily T, member 2, Sequence like an intermediate conductance potassium channel subunit, SLICKMGC119613, SLO2.1, sodium activated potassium channel, sodium and chloride activated ATP sensitive potassium channel, sodium- and chloride-activated ATP-sensitive potassium channel, Sodium and chloride-activated ATP-sensitive potassium channel Slo2.1, sodium-and chloride-activated ATP-sensitive potassium channel (SLICK)
Gene Symbols KCNT2
Host Species Mouse
Immunogen Fusion protein amino acids 564-624 of mouse Slo2.1 (Slick)
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 343450
Test Specificity Detects approx 140kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.