Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCP Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017039 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017039 Supplier Novus Biologicals Supplier No. NBP281728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCP Polyclonal specifically detects KCP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen KCP
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA
Formulation PBS
Gene Alias CRIM2, KCP1, kielin/chordin-like protein, NET67
Gene Symbols KCP
Host Species Rabbit
Immunogen KCP antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human KCP. The immunogen is located within amino acids 1320 - 1370 of KCP. Amino Acid Squence: RAAGYRARREANAR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375616
Test Specificity At least two isoforms of KCP are known to exist; this antibody will only detect the larger isoform. KCP antibody is predicted to not cross-react with CRIM1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.