Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCP Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB164190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164190 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164190 Supplier Novus Biologicals Supplier No. NBP281728JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCP Polyclonal antibody specifically detects KCP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen KCP
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CRIM2, KCP1, kielin/chordin-like protein, NET67
Host Species Rabbit
Immunogen KCP antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human KCP. The immunogen is located within amino acids 1320 - 1370 of KCP. Amino Acid Squence: RAAGYRARREANAR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375616
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.