Learn More
Invitrogen™ KCTD10 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Specifications
Specifications
| Antigen | KCTD10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | KCTD10 |
| Gene Accession No. | Q9H3F6 |
| Gene Alias | AW536343; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3; BTB/POZ domain-containing protein KCTD10; BTBD28; C87062; hBACURD3; KCTD10; mBACURD3; MSTP028; potassium channel tetramerisation domain containing 10; potassium channel tetramerization domain containing 10; potassium channel tetramerization domain-containing 10; potassium channel tetramerization domain-containing protein 10; ULR061; ULRO61; wu:fb30g12; zgc:63846 |
| Gene Symbols | KCTD10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.