Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD12 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096127 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096127 100 μL
NB096128 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096127 Supplier Novus Biologicals Supplier No. NBP304820100UL

Rabbit Polyclonal Antibody

KCTD12 Polyclonal specifically detects KCTD12 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen KCTD12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BTB/POZ domain-containing protein KCTD12, C13orf2, chromosome 13 open reading frame 2, FLJ33073, KIAA1778PFET1PFETIN, Pfetin, potassium channel tetramerisation domain containing 12, Predominantly fetal expressed T1 domain
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 196-325 of human KCTD12 (NP_612453.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115207
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.