Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD14 Antibody (OTI2E9), PE, Novus Biologicals™
SDP

Catalog No. NB288338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288338 Supplier Novus Biologicals Supplier No. NBP272171PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCTD14 Monoclonal antibody specifically detects KCTD14 in Human samples. It is validated for Western Blot

Specifications

Antigen KCTD14
Applications Western Blot
Classification Monoclonal
Clone OTI2E9
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias BTB/POZ domain-containing protein KCTD14, MGC2376, potassium channel tetramerisation domain containing 14
Host Species Mouse
Immunogen Full length human recombinant protein of human KCTD14 (NP_076419) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65987
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.