Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD15 Antibody (AT4C3), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N149429APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N149429APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N149429APC7 Supplier Novus Biologicals Supplier No. NBP149429APCCY7

Mouse Monoclonal Antibody

KCTD15 Monoclonal specifically detects KCTD15 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KCTD15
Applications Flow Cytometry
Classification Monoclonal
Clone AT4C3
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15
Gene Symbols KCTD15
Host Species Mouse
Immunogen Recombinant human KCTD15 (1-234aa) purified from E. coli.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79047.0
Test Specificity Human KCTD15 (1-234aa)
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG3 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.