Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD15 Antibody (AT4C3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP149429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149429 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149429 Supplier Novus Biologicals Supplier No. NBP149429
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCTD15 Monoclonal specifically detects KCTD15 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen KCTD15
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone 4C3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250-1:500, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:250
Gene Accession No. NP_076981
Gene Alias BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15
Gene Symbols KCTD15
Host Species Mouse
Immunogen Recombinant human KCTD15 (1-234aa) purified from E. coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79047
Test Specificity Human KCTD15 (1-234aa)
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG3 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.