Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCTD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568478
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA568478 100 μL
1 options

Catalog No. PIPA568478

Supplier: Invitrogen™ PA568478

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human and Rat samples.

The KCTD gene family, including KCTD7, encode predicted proteins that contain N-terminal domain that is homologous to the T1 domain in voltage-gated potassium channels (see KCNA1; MIM 176260). KCTD7 displays a primary sequence and hydropathy profile indicating intracytoplasmic localization. EST database analysis showed that KCTD7 is expressed in human and mouse brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCTD7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCTD7
Gene Accession No. B1WC97, Q8BJK1, Q96MP8
Gene Alias 4932409E18; 9430010P06Rik; BTB/POZ domain-containing protein KCTD7; CLN14; EPM3; FLJ32069; KCTD7; potassium channel tetramerisation domain containing 7; potassium channel tetramerization domain containing 7
Gene Symbols KCTD7
Host Species Rabbit
Immunogen Synthesized peptide derived from human KCTD7 [aa 181-230].
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154881, 212919, 688993
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.