Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDELR1 Antibody (KR-10), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP212873UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212873UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212873UV Supplier Novus Biologicals Supplier No. NBP212873UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KDELR1 Monoclonal specifically detects KDELR1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Hamster, Mammal, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen KDELR1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KR-10
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias ER lumen protein retaining receptor 1, ERD2, ERD2.1KDEL receptor 1, HDEL, KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1, KDEL endoplasmic reticulum protein retention receptor 1, PM23, Putative MAPK-activating protein PM23
Gene Symbols KDELR1
Host Species Mouse
Immunogen A 21 residue synthetic peptide (amino acids 192-212) based on the bovine KDEL receptor and the peptide coupled to KLH
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10945
Test Specificity Detects approx 25kDa.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Drosophila, Hamster, Mammalia, Monkey, Rabbit, Sheep, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.