Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM2A/FBXL11 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP178305UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178305UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178305UV Supplier Novus Biologicals Supplier No. NBP178305UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KDM2A/FBXL11 Polyclonal specifically detects KDM2A/FBXL11 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen KDM2A/FBXL11
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias [Histone-H3]-lysine-36 demethylase 1A, CXXC8F-box protein FBL7, CXXC-type zinc finger protein 8, FBL11DKFZp434M1735, FBL7F-box protein Lilina, F-box and leucine-rich repeat protein 11DKFZP434M1735, F-box protein FBL11, FBXL11FLJ46431, FLJ00115, JHDM1AF-box/LRR-repeat protein 11, JmjC domain-containing histone demethylation protein 1A, jumonji C domain-containing histone demethylase 1A, KIAA1004EC 1.14.11.27, LILINA, lysine (K)-specific demethylase 2A, lysine-specific demethylase 2A, SF-KDM2A
Gene Symbols KDM2A
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human KDM2A/FBXL11 protein (between residues 350-500) [UniProt Q9Y2K7]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 22992.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.