Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM2A/FBXL11 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166250 20 μg
NB166249 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166250 Supplier Novus Biologicals Supplier No. NBP31557720UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KDM2A/FBXL11 Polyclonal antibody specifically detects KDM2A/FBXL11 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen KDM2A/FBXL11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias [Histone-H3]-lysine-36 demethylase 1A, CXXC8F-box protein FBL7, CXXC-type zinc finger protein 8, FBL11DKFZp434M1735, FBL7F-box protein Lilina, F-box and leucine-rich repeat protein 11DKFZP434M1735, F-box protein FBL11, FBXL11FLJ46431, FLJ00115, JHDM1AF-box/LRR-repeat protein 11, JmjC domain-containing histone demethylation protein 1A, jumonji C domain-containing histone demethylase 1A, KIAA1004EC 1.14.11.27, LILINA, lysine (K)-specific demethylase 2A, lysine-specific demethylase 2A, SF-KDM2A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KDM2A/FBXL11 (NP_036440.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 22992
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.