Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM2B Antibody, Novus Biologicals™
SDP

Catalog No. NB1001293 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001293 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001293 Supplier Novus Biologicals Supplier No. NB1001293
Add to Cart
Add to Cart

Goat Polyclonal Antibody

KDM2B Polyclonal specifically detects KDM2B in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen KDM2B
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q8NHM5
Gene Alias [Histone-H3]-lysine-36 demethylase 1B, CXXC2F-box protein FBL10, CXXC-type zinc finger protein 2, Fbl10, F-box and leucine-rich repeat protein 10JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein, FBXL10, JHDM1BF-box/LRR-repeat protein 10, JmjC domain-containing histone demethylation protein 1B, jumonji C domain-containing histone demethylase 1B, Jumonji domain-containing EMSY-interactor methyltransferase motif protein, lysine (K)-specific demethylase 2B, lysine-specific demethylase 2B, PCCX2EC 1.14.11.27, protein containing CXXC domain 2, Protein JEMMA, Protein-containing CXXC domain 2
Gene Symbols KDM2B
Host Species Goat
Immunogen Peptide with sequence C-FGQVEEKLLQKLS corresponding to C-Terminus according to NP_115979.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84678
Test Specificity This antibody is expected to recognize isoform a (NP_115979.3) only.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.