Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM6A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092768 20 μL
NB092767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092768 Supplier Novus Biologicals Supplier No. NBP30510320UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KDM6A Polyclonal specifically detects KDM6A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen KDM6A
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias bA386N14.2, bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeatprotein (UTX)), DKFZp686A03225, EC 1.14.11, EC 1.14.11.-, Histone demethylase UTX, lysine (K)-specific demethylase 6A, MGC141941, ubiquitously transcribed tetratricopeptide repeat protein X-linked, ubiquitously transcribed tetratricopeptide repeat, X chromosome, ubiquitously transcribed TPR protein on the X chromosome, ubiquitously transcribed X chromosome tetratricopeptide repeat protein, ubiquitously-transcribed TPR gene on the X chromosome, Ubiquitously-transcribed TPR protein on the X chromosome, Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein, UTXlysine-specific demethylase 6A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 550-700 of mouse KDM6A (NP_033509.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7403
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.