Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kelch-Like Family Member 21 Antibody (2H4), Novus Biologicals™

Mouse Monoclonal Antibody
$487.50
Specifications
Antigen | Kelch-Like Family Member 21 |
---|---|
Clone | 2H4 |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Kelch-Like Family Member 21 Monoclonal specifically detects Kelch-Like Family Member 21 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kelch-Like Family Member 21 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
Ascites with 0.03% Sodium Azide | |
9903 | |
Purified recombinant fragment of human KLHL21 expressed in E. Coli. | |
Primary | |
66 kDa |
2H4 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
kelch-like 21 (Drosophila), Kelch-Like Protein 21, KIAA0469, KLHL21 | |
KLHL21 | |
IgG1 | |
Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title