Learn More
Stoelting™ Kelly Forceps
SDP
Kelly Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
$159.00 - $299.50
Specifications
For Use With (Application) Neuroscience
Length (English) 5.5 in.
View More Specs
Products 4
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-001-332
SDP
View Documents
Stoelting™
5211014
Straight
Each for $159.00
Only null left
Add to Cart
10-001-333
SDP
View Documents
Stoelting™
5211015
Straight, Left-handed
Each for $299.50
Only null left
Add to Cart
10-001-334
SDP
View Documents
Stoelting™
5211016
Curved
Each for $159.00
Only null left
Add to Cart
10-001-335
SDP
View Documents
Stoelting™
5211017
Curved, Left-handed
Each for $293.00
Only null left
Add to Cart
Description
Description
  • Scissor-like ring handles just like scissors
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Specifications
Specifications
Neuroscience
5.5 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.