Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kemtec™ ABO Blood Simutype Kit
Shop All AquaPhoenix Scientific Products

Click to view available options
Grade:
8 to 14
Description
Includes
- Microscope slides
- Simulated Anti-A antiserum
- Simulated Anti-B antiserum
- Simulated blood samples from four individuals
- 50 Hema-tags
- Green and yellow toothpicks
- Instructor's manual
- Reproducible student instructions and data sheet
- Convenient self-adhesive Hema-Tags™ typing templates
- Materials for up to 225 individual tests
- SDS
Blood typing has always been an important biology exercise because it introduces a number of key subjects. But the concern over AIDS, plus the general trend away from “invasive” laboratory work, has caused many schools to drift away from including this vital exercise. This kit provides a solution to all of these problems.
- Students use genetic information to answer paternity questions, determine genotypes and phenotypes of blood donors, and determine possible donors and recipients for blood transfusions
- Ideal for biology, immunology, genetics, or forensics
- Grades 7-12
- Class size: 24 students (with multiple reuses)
Specifications
Specifications
| Grade | 8 to 14 |
| For Use With (Application) | To determine possible blood donors for each blood type |
| Includes | One anti-a serum 5ml, one anti-b serum 5ml, four 2ml blood samples (simulated), 50 hema-tags, 12 microscope slides, 50 blue, 50 yellow toothpicks , 50 student instructions, instructor's manual, teacher's guide, worksheets for 50 students |
| Class Size | 50 Students |
| Product Type | Abo Blood Simutype Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction