Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ ABO Blood Typing Kit (with Red Blood Cells)
SCI_ED

Catalog No. s17020 Shop All AquaPhoenix Scientific Products
Click to view available options
Grade:
8 to 14
Choking hazard (1) Small parts. Not for children under 3 yrs.

Catalog No. S17020

Supplier: Aqua Phoenix Scientific 3196

Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Anti-A Serum, Anti-B Serum, Anti-Rh Serum, A Blood Cells, B Blood Cells, O Blood Cells, droppers, instructor manual, student data sheets, MSDS

Learn the genetic inheritance patterns of blood types.

This kit includes preserved red blood cells and eliminates the necessity of drawing blood samples from students. Kit should be ordered two weeks before needed.

 

  • Contains both red blood cells and antiserum needed to demonstrate real ABO blood typing agglutination reactions
  • Students use real red blood cells provided, eliminating the need to draw blood samples
  • Perform, compare, and contrast slide and test tube blood typing
  • Red cells have a life expectancy of at least 20 days after arrival, but must be kept under refrigeration
  • Grades 9-14
  • Class size: 36 students

 

Specifications

Grade 8 to 14
For Use With (Application) Eliminates the need for drawing blood samples from students
Includes One vial anti-a serum, one vial anti-b serum, four vials red blood cells (a, b, o and ab), one general history sheet one “what type blood are you?” sheet
Product Type Blood Typing Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.