Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Determining Molar Mass by Colligative Properties
SCI_ED

Catalog No. s07336 Shop All AquaPhoenix Scientific Products
Click to view available options
Grade:
10 to 14
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Sucrose, 50g; D-Glucose, 40g; Calcium Chloride, Anhydrous, 20g; Lauric Acid, 100g; Benzoic Acid, 14g; Sodium Chloride, Rock Salt; Naphthalene, 15g

Requires

Hotplates, analytical balances, thermometers and basic lab supplies

Students investigate all aspects of freezing point depression, including solvent freezing constant, van't Hoff factor and molar mass.

Fulfill the AP required inquiry component while determining important applications of freezing point depression in everyday life. Direct students to design an experiment to resolve: What is the lowest temperature at which road treatment containing calcium chloride would be effective?

  • Includes multiple solvents and solutes, both ionic and molecular
  • Instructor's manual features background material, easy-to-follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 2, 3 & 5
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science practices 2, 3 and 5
Product Type Determining Molar Mass By Colligative Properties Chemistry Kit
Grade 10 to 14
For Use With (Application) For students to investigate all aspects of freezing point depression, including solvent freezing constant, van't hoff factor and molar mass
Includes 50g sucrose, 40g d-glucose, 20g calcium chloride anhydrous, 100g lauric acid, 14g benzoic acid, sodium chloride, 15g rock salt, naphthalene
Class Size 24 Students (12 Groups)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.