Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Le Chatelier's Principle Featuring Ag+ reaction!
SCI_ED

Catalog No. S07340 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
24 Students (12 Groups)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S07340 24 Students (12 Groups)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07340 Supplier Aqua Phoenix Scientific Supplier No. 16113
Only null left
Add to Cart
Add to Cart

Includes

Sulfuric Acid, 50%v/v (1:1), 15mL; Hydrochloric Acid, 12M, 4mL; Hydrochloric Acid, 50% v/v, 1:1; Sodium Hydroxide, 6M; Silver Nitrate, 2M, 4mL; Aluminum Potassium Sulfate, 400g; Cobalt Chloride Hexahydrate, 25g; Sodium Chloride, 650g; Methyl Red, 0.1% w/v, 4mL; Bromthymol Blue Ind, 4mL; Sodium Nitrate, ACS, 25g

Requires

Standard lab equipment and fume hood

Students create and analyze four different equilibrium systems, allowing manipulation of a variety of stresses to show how systems respond by shifting in the direction that reduces stress.

Students vary temperature, alter the pH of a weak acid indicator, use silver ions to draw chloride from a complex ion by precipitation, and design a set of conditions for optimum yield. Students make predictions and test their hypotheses.

  • Ideal for AP Chemistry, Chemistry II or Honors Chemistry
  • Instructor's manual features background material, easy to follow stepwise protocols, assessment questions, sample data, and an answer key
  • Experiments are readily adjustable to fit your lab periods
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 6 & 7
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science practices 6 and 7
Grade 10 to 14
For Use With (Application) For ap chemistry, chemistry ii or honors chemistry
Includes 50% v/v (1:1), 15mL Sulfuric Acid, 12M, 4mL Hydrochloric Acid, 50% v/v, 1:1 Hydrochloric Acid, 6M Sodium Hydroxide, 2M, 4mL Silver Nitrate, 400g Aluminum Potassium Sulfate, 25g Cobalt Chloride Hexahydrate, 650g Sodium Chloride, 0.1% w/v, 4mL Methyl Red, 4mL Bromthymol Blue Ind, ACS, 25g Sodium Nitrate
Class Size 24 Students (12 Groups)
Product Type Le Chatelier'S Principle Featuring Ag+ Reaction! Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.