Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP Chemistry Kits: Preparing Buffer Solutions and Determining Their Properties
SCI_ED

Catalog No. S07342 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Preparing Buffer Solutions and Determining Their Properties
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S07342 Preparing Buffer Solutions and Determining Their Properties
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07342 Supplier Aqua Phoenix Scientific Supplier No. 16116

missing translation for 'validMainframeMsg'

Includes

Sodium Hydroxide, 12.0N; Hydrochloric Acid, 12M; Acetic Acid, 25%v/v, 100ML; Ammonium Hydroxide, 20%v/v 100mL; Sodium Phosphate Monobasic; Sodium Phosphate Dibasic; Ammonium Chloride, ACS; Sodium Acetate, Trihydrate, ACS

Requires

pH meter, glassware

After being assigned one of the three buffer systems, pH, and component acids, bases or conjugates, students construct a buffer and test it based on their own calculations. Students compare the pH changes of their buffer to those of distilled water and other solutions.

  • Minimal prep time required
  • Instructor's manual features background material, easy to follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 4 & 7
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science practices 4 and 7
Grade 10 to 14
For Use With (Application) For students construct a buffer and test it based on their own calculations, students compare the ph changes of their buffer to those of distilled water and other solutions
Includes 12n sodium hydroxide, 12m hydrochloric acid, 25%v/v, 100ml acetic acid, 20%v/v 100ml ammonium hydroxide, sodium phosphate monobasic, sodium phosphate dibasic, ammonium chloride, acs, sodium acetate, trihydrate, acs
Class Size 24 Students (12 Groups)
Product Type Preparing Buffer Solutions and Determining their Properties Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.