Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Spectrophotometric Analysis of Copper: Beer's Law
SCI_ED

Catalog No. S07330 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
10 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07330 10 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07330 Supplier Aqua Phoenix Scientific Supplier No. 16102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Copper Sulfate, Pentahydrate; Copper Sulfate 0.12M; Copper Sulfate 0.012M; Copper Sulfate 0.0024M; Copper Sulfate 0.00048M

Requires

Spectrophotometers, 10mL volumetric flasks, 1mL pipets

Explore the interaction of light with copper ions in solution using photons from the visible portion of the electromagnetic spectrum.

Students examine the relationship between transmittance, absorbance and solution concentration using Beer's Law (A = ε·b·C) after determining λmax. Fulfill the AP chemistry requirement for using a spectrophotometer and learn to perform serial dilutions.

  • Includes an inquiry based extension for determining the amount of copper in a penny (nitric acid not included)
  • Experiments are adjustable to fit your lab period
  • Minimal prep time
  • Ideal for AP or Honors Chemistry
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Instructor's manual features background material, easy to follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Meets AP Science Practices: 2, 3 & 5
  • Targets Grades: 10 to14
  • Accommodates 24 Students Working in Groups of 2

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire™ handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets AP Science Practice 2, exceed 2013 AP Next Generation National Standards
Grade 10 to 14
For Use With (Application) For students examine the relationship between transmittance, absorbance and solution concentration using beer's law (a = e.b.c) after determining lmax, for ap or honors chemistry
Includes Copper sulfate, pentahydrate, 0.12m copper sulfate, 0.012m copper sulfate, 0.0024m copper sulfate, 0.00048m copper sulfate
Class Size 24 Students (12 Groups)
Product Type Spectrophotometric Analysis of Copper, Beer'S Law Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.