Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Stoichiometry: Mole Ratio of an Unknown
SCI_ED

Catalog No. S07335 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
10 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07335 10 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07335 Supplier Aqua Phoenix Scientific Supplier No. 16107
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Two bottles Citric Acid Anhydrous; two bottles Sodium Hydroxide, 5.0N, 200mL

Requires

Glassware, thermometers and styrofoam cups

This unique, hands-on lab shows students how to treat heat as a quantitative product, using continuous variation (Job's method).

The reagent combination used provides reliable results.

  • Instructor's manual features background material, easy-to-follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Minimal prep time
  • Easily adapted to shorter lab periods
  • Suitable for Honors and AP chemistry
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 2, 3 & 5
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science practices 2, 3 and 5
Grade 10 to 14
For Use With (Application) To show students how to treat heat as a quantitative product, using continuous variation (job's method)
Includes Two bottles citric acid anhydrous, 5.0n, 200ml two bottles sodium hydroxide
Class Size 24 Students (12 Groups)
Product Type Mole Ratio of An Unknown Stoichiometry Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.