Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Build Your Own Simple Electric Motor Kit
SCI_ED

Catalog No. S07349 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
5 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07349 5 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S07349 Supplier Aqua Phoenix Scientific Supplier No. 15210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Magnet wire, copper wire, rectangular magnets, D-Cell battery holder w/leads, round magnets, small nails, ring magnet, wire cutter, neodymium magnet, sandpaper, dowel rods, instructor's manual

Requires

Safety goggles and batteries (three D-cell for single-pack kit and 18 D-cell for class-pack kit).

Build your own simple electric motors using wire and magnets.

  • Hands-on project set designed for whole class to extend learning from electromagnets to motors
  • Great for introducing guided inquiry-based learning, systematic problem solving and teaching engineering design practices
  • Explore effects of altering coils in motor, wire thickness and effects of using different sizes, shapes, strengths and types of magnets
  • Includes instructor's manual with reproducible stepwise student protocols
  • Includes four experiments which culminate in a design activity
  • With instructor's manuals with lesson plans, background information, reproducible stepwise student protocols and guided worksheets
  • Availalbe as single kit (1 to 4 students) or class pack kit (24 students)
  • Grade 5 to 8

Specifications

Grade 5 to 8
For Use With (Application) Designed for whole class to extend learning from electromagnets to motors
Includes 4 experiments, background information, sample data and complete answer keys, instructor's manual
Product Type Physics Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.