Learn More
Description
Includes
Instructor manual, student manuals, fishline, spacers, washers, rubber stoppers, paper clips, alligator clips, metric tape, stopwatch
- Explore tangential motion and centripetal acceleration
- Students record real data to calculate real forces
- Instructor demonstration included
- Instructor's manual includes guided calculations, assessment questions, sample data and answer keys
- Appropriate for any level physics course (including AP) as well as general science courses
- Grades 9 to 12
- Class size: 24 students
Order Info
Impact goggles and a balance accurate to 0.01g.
Specifications
Specifications
| Grade | 8 to 12 |
| For Use With (Application) | Illustrates the experience of centrifugal force and its opposite, centripetal force in a simple experiment |
| Includes | Instructor manual, student manuals, fishline, spacers, washers, rubber stoppers, paper clips, alligator clips, metric tape, stopwatch |
| Class Size | 36 Students |
| Product Type | Circular Motion Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
