Learn More
Kemtec™ Cryolizer Cold Chamber Kit
SCI_ED
Catalog No. s06749 Shop All AquaPhoenix Scientific Products
Click to view available options
Diameter (English) Outside:
3.5 in.
Catalog No. S06749
Supplier: Aqua Phoenix Scientific 2800
Only null left
Add to Cart
Add to Cart
Includes
Cryolizer Unit, Growth Chamber, Observation dish, Sheet of Instructions
Immobilization periods up to one hour without the use of chemicals.
  • Immobilization relies on the cold sensitivity of insects
  • Low temperature for extended period maintained by chamber that utilizes unique thermal transfer that acts as “cold sink”
  • For grades 6 to 12
Specifications
Material Plastic
Type Cryolizer Cold Chamber Kit
Includes Cryolizer Unit, observation dish, growth chamber, instructions
Height (English) 3.5 in.
Diameter (English) Outside 3.5 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.