Learn More
Kemtec™ DNA Model with Paint Kit
SCI_ED
Catalog No. s25825
Click to view available options
:
DNA molecular model
Includes
Instructor manual, student problem sheet, figure sheets (13 figures), metal rod paired wooden base, AT (Adenine-Thymine), GC (Guanine-Cytosine) deoxyribose units, abbreviated phosphate groups, carbon atoms (of deoxyribose) bond phosphate to carbon (of deoxyribose) bond, spacers, flat washer, threaded washer, wing nut, top nut, set of six different colors of model paint and paint thinner
Enhance lectures or labs with this model enlarged 100 million times (scale 10cm x 1nm).
  • The model depicts DNA structure containing 16 nucleotide pairs (eight AT, eight GC pairs)
  • Fast and easy assembly with Pop-it™ bead construction
  • Model is shipped unassembled and unpainted with chrome-like stand and stained mahogany base
  • Dimensions: 60.9 x 20.3cm (24 x 8 in.)
  • Grades 7 to 14
  • Building and painting project ideal for six students at time
Demonstrates:
  • Antiparallel DNA strands
  • Ten nucleotide pairs/revolution
  • Five prime and three prime ends of strands
  • Hydrogen bonding of basepairs AT vs. GC base pairing
  • 0.34nm spacing
  • 3.4nm pitch
Painting is not required for normal use.
Specifications
Product Type DNA Model with Paint
For Use With (Application) Demonstrate antiparallel DNA strands, ten Nucleotide pairs/revolution, five prime and three prime ends of strands, Hydrogen bonding of basepairs AT vs. GC base pairing 0.34nm spacing 3.4nm pitch
Includes Instructor manual, student problem sheet, figure sheets (13 figures), metal rod paired Wooden base, AT (Adenine-Thymine), GC (Guanine-Cytosine) deoxyribose units, abbreviated phosphate groups, Carbon atoms (of deoxyribose) bond phosphate to Carbon (of deoxyribose) bond, spacers, flat washer, threaded washer, wing nut, top nut, set of six different colors of model paint and paint thinner
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.