Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Food Chain Kit
SCI_ED

Catalog No. S02091 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Food Chain Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S02091 Food Chain Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S02091 Supplier Aqua Phoenix Scientific Supplier No. 1124

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

Instructor manual, student data sheets, overhead transparencies, food chain picture card game, CD with images, Ziploc™ bags and cotton swabs

Introduces students to the concepts of food chains.

  • For students to study different relationships within an environment, importance of each organism, how energy is transferred, and delicate balance in environment of food web
  • Discuss producers, consumers, herbivores, carnivores, omnivores, predators, prey, decomposition, energy flow, food cycles, food chains, food webs, and food pyramids
  • Observe pond water and soil habitat food webs and pyramids
  • Study effects of acid rain, road salt and field runoff on environments
  • Model decomposition of plant matter
  • Design own food webs
  • Grades 4-7
  • Class size: 24 students

Specifications

Grade 4 to 6
Product Type Food Chain Kit
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.