Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Forensic Blood Spatter Kit
SCI_ED

Catalog No. s68542 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
:
Blood Spatter Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S68542 Blood Spatter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S68542 Supplier Aqua Phoenix Scientific Supplier No. 10550
Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Instructor's manual, assessment manual, MSDS, student data sheets, Sixteen General Rules of Blood Spatter, Thirty Seven Investigative Terminology Terms, blood spatter test, large paper clip, pipets, magnifying glasses, protractors, measuring tape, filter paper, rubber bands, clipboards, cardboard, spray bottles, labels, blood spatter chamber, dowel rods, hammer, striking platform, paper for spatter, simulated blood solution, simulated hemoglobin solution, luminol powder, sodium perborate, sodium carbonate

Requires

Newspaper, cleanup towels, water, black light, masking tape, UV light

Examines how blood spatter plays a role in crime scene investigation.

Students determine how the height and angle of a blood drop impact the size and shape of a blood spatter, then use this information along with the sixteen general rules of spatter interpretation to analyze data.

  • Experiments introduce techniques used by forensic specialists
  • Test and understand basic principles and terminology
  • Grades: 9 to 14
  • Class size: 24 students

Lessons include:

  • Distance effect on blood spatter
  • Identifying distance and angle of travel with blood spatter
  • Investigating blood spatter with luminol
  • Assembling a blood spatter chamber and experimenting with spatter (includes the 16 general rules of blood spatter interpretation and 37 investigative terminology terms)
  • Creating a crime scene scenario

Specifications

Certifications/Compliance National standards
Grade 8 to 14
For Use With (Application) Examines how blood spatter plays a role in crime scene investigation
Includes Instructor manual, student data sheets for 5 experiments, MSDS, sixteen general rules of Blood spatter, thirty seven investigative terminology terms, Blood spatter test, large Paper clip, pipets, magnifying Glasses, protractors, measuring tape, filter Paper, Rubber bands, clip boards, clip boards, spray bottles, labels, Blood spatter chamber, dowel rods, hammer, striking platform, Paper for spatter, simulated Blood solution, simulated hemoglobin solution, luminol powder, Sodium Perborate, Sodium Carbonate
Requires Black light
Class Size 24 Students
Product Type Blood Spatter Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.