Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Human Activity and the Environment Kit
SCI_ED

Catalog No. S02076 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
1 to 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02076 1 to 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S02076 Supplier Aqua Phoenix Scientific Supplier No. 1169
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Includes

Instructor manual, transparencies How the World Works, photocopy sheets for activities: crossword, Forest Community, Runoff, Builder A&B, Flooded Farms, Salted Roads and true/false questions. Also, fact sheets, bean seeds, plastic cups, Miracle-Gro™, vinegar, methylene blue stain and sand.

Requires

Fresh celery, dominoes, recycled milk jugs or flower pots, beakers or jars, yeast, salt, and sugar.

Helps students understand how human activity affects the environment.

Students study food web relationships and how natural and human induced changes in the environment such as severe storms, drought, floods, housing developments, road salt, fertilizer runoff, and fire upset the balance of ecosystems.

  • Demonstrates nature's ability to restore balance between number of different types of organisms, such as predators and prey, in natural habits of community
  • Activities include: The Salted Roads, The Runoff, Flooded Farms and The Forest Community
  • Conservation is discussed in terms of using natural resources wisely to help keep natural balance
  • Grades 5-8
  • Class size: 24 students

Specifications

Grade 1 to 6
Product Type Human Activity and The Environment Kit
Includes Instructor manual, transparencies How the World Works, photocopy sheets for activities: crossword, Forest Community, Runoff, Builder A and B, Flooded Farms, Salted Roads and true/false questions. Also, fact sheets, bean seeds, Plastic cups, Miracle-Gro™
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.