Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Let's Build Electromagnets Kit
SCI_ED

Catalog No. s07348 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
3 to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07348 3 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Choking hazard (1) Small parts. Not for children under 3 yrs.

Catalog No. S07348 Supplier Aqua Phoenix Scientific Supplier No. 15205
Only null left
Add to Cart
Add to Cart

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Dowel rod, alligator clip leads, 6V lantern battery, wire strippers, magnet wire, masking tape, paper clips, steel washers, steel nails, sandpaper, steel bolts, weigh dish, instructor's manual, reproducible student data sheets

Requires

Impact goggles

Introduce the Engineering Design Process for problem solving as you explore electricity and magnetism in this electromagnet building exercise.

  • Explore magnetic field, electrical charge, and effects of increasing current on the generation of heat
  • Compare core materials, core thickness, and number of wraps in coils in inquiry based investigations
  • Four engaging activities culminate with students applying what they learn in a design the strongest electromagnet activity
  • Instructor's manual with lesson plans, background information, reproducible stepwise student instructions, worksheets, and complete answer keys
  • Grades 3 to 5
  • Available as single kit (1 to 4 students) or class pack kit (24 students)

Specifications

For Use With (Application) Introduce students to electricity and magnetism with this electromagnet building exercise
Grade 3 to 5
Product Type Class Pack Kit
Includes Instructor's manual, student guide, worksheets, batteries and all materials required for investigation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.