Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Make an Ocean Kit
SCI_ED

Catalog No. s02088 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
4 to 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02088 4 to 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
CHOKING HAZARD—Small parts. Not for children under 3 yr.
Catalog No. S02088 Supplier Aqua Phoenix Scientific Supplier No. 1140
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
CHOKING HAZARD—Small parts. Not for children under 3 yr.

Includes

Instructor manual, life-cycle chart, ocean mat, medicine cup, hydrometer, pH indicator paper, measuring spoon, hand lens, soda straw, pipette, toothpicks, labels for types of waste, slow-motion fluid, brine shrimp eggs, brine shrimp food, farm runoff crystals, industrial waste crystals, chemical treatment and sea salt.

Requires

Five one gallon containers, balance and a flashlight.

Students model four ocean ecology scenarios as they investigate the effect of farm and industrial waste on ocean life.

  • Populations of brine shrimp are monitored over the course of ecological contamination, waste treatment and recovery
  • Students learn about life cycles and representative sampling
  • Long-term experiment ideal for class project or small groups
  • Grades 4-6
  • Class size: 24 students

Specifications

Grade 4 to 6
For Use With (Application) Learn about life cycles and representative sampling
Includes Instructor manual, life-cycle chart, ocean mat, medicine cup, hydrometer, pH indicator paper, measuring spoon, hand lens, soda straw, pipet, toothpicks, labels for types of waste, slow-motion fluid, brine shrimp eggs, brine shrimp food, farm runoff crystals, industrial waste crystals, chemical treatment and sea salt
Class Size 24 Students
Description Make an Ocean Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.