Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Ocean Science Kit
SCI_ED

Catalog No. s94962 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
5 to 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S94962 5 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S94962 Supplier Aqua Phoenix Scientific Supplier No. 9118
Only null left
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Instructor Manual, Student Data Sheets, Graph Paper, Petri Dishes, Food Coloring, Sea Salt, Hydrometer, Plastic Cups

Requires

Ice, a refrigerator/freezer, thermometers and 18 large beakers or jars

Examine physical and chemical properties of seawater and learn how these properties affect ocean currents and environments.

Students determine the effects of three scenarios on the surrounding water and land environments and inhabitants: 1) a new dam being built on a large river that flows into a bay, 2) melting of the polar ice cap, and 3) a global temperature drop to 0°C and present their conclusions in a panel discussion.

  • Visualize convection currents while tracking effects of density, temperature, pressure and salinity
  • Learn effects of sun heat, precipitation, evaporation, freezing and thawing of sea ice and influx of fresh water from rivers
  • Link science and technology to local, national, and global changes
  • Grades 5-9
  • Class size: 24 students

Specifications

Grade 5 to 9
For Use With (Application) Examine physical and chemical properties of seawater and learn how these properties affect ocean currents and environments
Includes Instructor Manual, Student Data Sheets, Graph Paper, Petri Dishes, Food Coloring, Sea Salt, Hydrometer, Plastic Cups
Class Size 24 Students
Description Ocean Science Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.