Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kemtec™ Osmosis and Diffusion Kit PROMO
Shop All AquaPhoenix Scientific Products

Click to view available options
Grade:
8 to 12
Description
Students will utilize starch and glucose solutions with dialysis tubing to model the cell membrane and observe the processes of osmosis and diffusion (essential concepts in understanding how cells survive).
- Students can experiment with membranes and molecules
- Illustrates cellular absorption in animal tissue
- Helps to visualize exchange between water and solutes
- Kit contains materials to carry out two complete experiments with glucose and starch
- Three part lab takes no more than two class periods to complete
- This laboratory activity is ideal for high school biology classes of all levels
- For grades 9 to 12
- Class size: 24 students
Specifications
Specifications
Grade | 8 to 12 |
Includes | Instructor manual, student data sheets, methods of assessment manual, material safety data sheets, glucose test strips, dialysis tubing, pipets, two dram vials, iodine solution, glucose, starch |
Product Type | Osmosis and Diffusion Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction