Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Photosynthesis I (Hill Reaction) Kit
SCI_ED

Catalog No. s06747 Shop All AquaPhoenix Scientific Products
Click to view available options
:
Photosynthesis I (Hill Reaction) kit
This item is not returnable. View return policy
Choking hazard (1) Small parts. Not for children under 3 yrs.
This item is not returnable. View return policy
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

Sucrose, dichlorophenol (DPIP), sodium hydrosulfite, propylene glycol, potassium phosphate dibasic, potassium phosphate monobasic, potassium chloride, instructor manual, student data sheets, methods of assessment manual, SDS

Requires

Centrifuge, blender, spectrophotometer (optional), light source, glassware, fresh spinach or lettuce leaves.

Use this kit to perform partial processes of photosynthesis in the laboratory.

Green plant photosynthesis is an oxidation reduction process in which hydrogen is split for water and used to reduce carbon dioxide to the level of a carbohydrate. This proteolysis of water when carried out with isolated chloroplasts is known as the Hill reaction after R. Hill who discovered it in 1937. It is necessary to supply a hydrogen acceptor in carrying out the Hill reaction as isolated chloroplasts do not reduce carbon dioxide directly.

  • Splitting of water with evolution of oxygen and release of hydrogen is measurable in course of the reaction by bleaching of hydrogen acceptor using a spectrophotometer or visual examination
  • This reaction is run in light and in dark to demonstrate light dependence of the reaction
  • For grades 9 to 14
  • Class size: 24 students

Specifications

Product Type Kemtec™ Photosynthesis I (Hill Reaction) Kit
Grade 8 to 14
For Use With (Application) Perform partial process of photosynthesis in the laboratory, provides hydrogen acceptor for proteolysis of water carried out with isolated chloroplasts
Includes Student data sheets, methods of assessment manual, material safety data sheet, sucrose, dichlorophenol (dpip), sodium hydrosulfite, propylene glycol, potassium phosphate dibasic, potassium phosphate monobasic, potassium chloride, instructor manual
Requires Centrifuge, blender, spectrophotometer (optional), a light source, glassware and fresh spinach or lettuce leaves
Class Size 24 Students
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.