Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ Resources of the Living Ocean
SCI_ED

Catalog No. S94961 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S94961 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Choking hazard (1) Small parts. Not for children under 3 yrs.
Catalog No. S94961 Supplier Aqua Phoenix Scientific Supplier No. 9122
Add to Cart
Add to Cart
Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Instructor Manual
  • MSDS
  • Student Data Sheets
  • Graph Paper
  • White Paper
  • Microscope Slides
  • Forceps
  • Rulers
  • Magnifying Glasses
  • Straws
  • Sodium Bicarbonate

Requires

Microscopes, fresh whole fish, elodea, lamps or sockets with bulbs, a cooler with ice and glassware

Designed to show importance of fish and algae as living resources of ocean.

  • Study relationship of form and function in fish by examining fin shapes and sizes, body shapes, mouth size and shape, color and camouflage
  • Predict habitats and behaviors by observing structures on fresh fish
  • Use dichotomous key to identify fish by genus species
  • Study oxygen production through photosynthesis in elodea
  • Grades 9-12
  • Class size: 24 students

Specifications

Grade 9 to 12
For Use With (Application) Shows importance of fish and algae as living resources of ocean
Includes Instructor Manual; MSDS; Student Data Sheets; Graph Paper; White Paper; Microscope Slides; Forceps; Rulers; Magnifying Glasses; Straws; Sodium Bicarbonate
Class Size 24 Students
Description Resources of the Living Ocean Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.