Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™: Robbery at Anytown Mall
SCI_ED

Catalog No. s06743 Shop All AquaPhoenix Scientific Products
Click to view available options
Grade:
7 to 11
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Cloth (simulated silk, cotton, linen, polyester, simulated wool), cat hair, human hair, dog hair, police envelope, suspect envelope, signature cards, hand lens, cover slips, microscope slides, wooden sticks, red litmus paper, tweezers, toothpicks, A and B hema blood tag, Rh hema blood tag, mounting media, anti-A blood simulated, anti-B blood simulated, anti-Rh blood simulated, type B simulated blood, type A simulated blood, type O simulated blood, type AB simulated blood, instructor manual, student manuals, MSDS, student data charts, special needs materials

Requires

Microscopes, water bath, and test tubes with stoppers

Students solve the case of a robbery of a designer linen suit as a CSI would.

  • Scientific methods employed by students for analysis are: observation, experimentation and interpretation
  • Both evidence and personal profiles are used to crack case using deductive reasoning
  • Grades 7-12
  • Class size: 24 students (six groups of four)
Eight lab activities included are:
  • Fabric Analysis
  • Fingerprint
  • Fiber Analysis
  • Hair Analysis
  • Handwriting
  • Analysis Blood Typing
  • Personal Information
  • Questions and Comments

Specifications

Certifications/Compliance National standards
Product Type Robbery At Anytown Mall Lab Activity Kit
Grade 7 to 11
For Use With (Application) For students solve the case of a robbery of a designer linen suit as a csi would
Includes 8 labs for students are fabric analysis, fingerprint, fiber analysis, hair analysis, handwriting, analysis blood typing, personal information, questions and comments
Requires Microscope, water bath, and glass test tubes with stoppers or caps
Class Size 24 Students
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.