Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin 31 Antibody (LHTric-1), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309167 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309167 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309167 Supplier Novus Biologicals Supplier No. NBP253114MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keratin 31 Monoclonal antibody specifically detects Keratin 31 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Keratin 31
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone LHTric-1
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias Hair Keratin, Type I Ha, Hard Keratin, Type I, 1, hHa1, K31, Keratin 31, Keratin, Hair, Acidic, 1, Keratin, Type I Cuticular Ha1, Keratin-31, KRTHA1
Host Species Mouse
Immunogen CVPRPRCGPCNSFVR
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3881
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.