Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin, Powder, Spectrum™ Chemical
SDP

Catalog No. 18603635 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail

K3030

Spectrum™ Chemical Keratin, Powder is the key material that makes up the human body’s out layer of skin. It is also the key element of hair and nails. It is considered to be similar to chitin. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 69430-36-0
Molecular Formula C2H2BrClO2
Molecular Weight (g/mol) 173.39
InChI Key GEHJBWKLJVFKPS-UHFFFAOYNA-N
IUPAC Name 2-bromo-2-chloroacetic acid
SMILES OC(=O)C(Cl)Br

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 2.5 kg
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.