Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoconazole, 99.17%, MP Biomedicals™

Catalog No. ICN15915880
Click to view available options
Quantity:
50 mg
100 mg
500 mg

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
MDL Number MFCD00058579
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym cis-1-Acetyl-4-(4-((2-(2, 4-dichlorophenyl)-2-(1H-1M-idazol-1-ylmethyl)-1, 3-dioxolan-4-yl) methoxy) phenyl)-piperazine, Ketoisdin
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Type Ketoconazole
Physical Form Powder
Color White
Quantity 100 mg
Assay Percent Range >99%
Formula Weight 531.44
Chemical Name or Material Ketoconazole
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.