Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid Dihydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. A219250 Shop All Fisherbrand Products
Click to view available options
Quantity:
250 g
500 g
3 kg
50 kg
Packaging:
Poly Bottle
Poly Drum
This item is not returnable. View return policy

Catalog No. A219-250

Only null left
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
MDL Number MFCD00149102
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

pH 1.3
Melting Point 98°C
Color White
Assay Percent Range 99.5 to 102.5%
Linear Formula HO2CCO2H·2H2O
UN Number 2923
Formula Weight 126.07
Percent Purity 99.5 to 102.5%
Grade Certified ACS
Density 4.62
Quantity 250 g
Ignition Residue 0.01% max.
Packaging Poly Bottle
Physical Form Powder/Solid
Vapor Pressure 21.5mbar at 50°C
Chemical Name or Material Oxalic Acid Dihydrate
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe eye burns. Harmful in contact with skin and if swallowed. Irritating to respiratory system. Corrosive to metals. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:1
Instability:0
DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.