Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boric Acid (Powder/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
Specifications
Specifications
pH | 3.8 to 4.8 |
Color | White |
Melting Point | 169°C |
Heavy Metals (as Pb) | 0.001% max. |
Identification | Pass Test |
Non_volatile Matter | 0.05% max. |
Packaging | Poly Bottle |
Vapor Pressure | 2.7mbar at 20°C |
Assay Percent Range | ≥99.5 % |
Quantity | 1 kg |
Show More |
Safety and Handling

Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview May impair fertility. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Ingestion may cause gastrointestinal irritation, nausea, vomiting and diarrhea. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:2 Flammability:0 Instability:1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction