Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetal, 99%

Catalog No. AC102222500
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 105-57-7 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-57-7
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009243
InChI Key DHKHKXVYLBGOIT-UHFFFAOYSA-N
Synonym acetal, acetaldehyde diethyl acetal, diethyl acetal, ethane, 1,1-diethoxy, diaethylacetal, acetale, ethylidene diethyl ether, acetal diethylique, diethylacetal, 1,1-dietossietano
PubChem CID 7765
IUPAC Name 1,1-diethoxyethane
SMILES CCOC(C)OCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless
Density 0.8250g/mL
Boiling Point 103.0°C
Flash Point −21°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3790 to 1.3850
Linear Formula CH3CH(OC2H5)2
Quantity 250 mL
Beilstein 01, 603
Specific Gravity 0.825
Merck Index 15, 39
Solubility Information Solubility in water: 46g/L (25°C). Other solubilities: miscible with alcohol,ether,soluble in heptane,methylcyclohexane,,ethyl acetate,propyl-,isopropyl-,butyl-,,isobutyl alcohol
Formula Weight 118.18
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Acetal, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-310-6
RTECSNumber AB2800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.