Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzyltriphenylphosphonium Chloride, 99%
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 337.0°C |
| Color | White |
| Flash Point | >300°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (Argentometry) |
| Packaging | Glass bottle |
| Linear Formula | C6H5CH2P(C6H5)3Cl |
| Quantity | 25 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF IN EYES: Rinse cautiously with wat |
| EINECSNumber | 214-154-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction