Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene, 99.8%, pure

Catalog No. AC118080025
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy

Catalog No. AC118080025

Supplier: Thermo Scientific Chemicals 118080025

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 100-41-4 | C8H10 | 106.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-41-4
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1

Specifications

Melting Point -95.0°C
Density 0.8600g/mL
Boiling Point 136.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 99.7% min. (GC)
Packaging Glass bottle
Molecular Formula C8H10
Refractive Index 1.4940 to 1.4970
Linear Formula C6H5C2H5
Quantity 2.5 L
Beilstein 05, II, 274
Specific Gravity 0.86
Merck Index 15, 3820
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, benzene, carbon tetrachloride,, ether and sulfur dioxide, slightly soluble in chloroform, insoluble in ammonia
Molecular Weight (g/mol) 106.17
Viscosity 0.63 mPa.s (20°C)
Formula Weight 106.17
Percent Purity 99.8%
Grade Pure
Physical Form Liquid
Chemical Name or Material Ethylbenzene, 99.8%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasti
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a
EINECSNumber 202-849-4
RTECSNumber DA0700000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.