Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione, 98%, for analysis, reduced

Catalog No. AC120000250
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
100 g
500 g
Catalog No. Quantity
AC120000250 25 g
AC120000010 1 g
AC120000050 5 g
AC120001000 100 g
AC120005000 500 g
5 missing translation for 'options'
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Catalog No. AC120000250

Supplier: Thermo Scientific Chemicals 120000250

Only null left
Add to cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

CAS: 70-18-8 | C10H17N3O6S | 307.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-18-8
Molecular Formula C10H17N3O6S
Molecular Weight (g/mol) 307.32
MDL Number MFCD00065939
InChI Key RWSXRVCMGQZWBV-WDSKDSINSA-N
Synonym glutathione, l-glutathione, glutathion, glutathione-sh, glutinal, isethion, tathion, reduced glutathione, deltathione, neuthion
PubChem CID 124886
ChEBI CHEBI:16856
IUPAC Name (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-1-oxo-3-sulfanylpropan-2-yl]amino]-5-oxopentanoic acid
SMILES C(CC(=O)NC(CS)C(=O)NCC(=O)O)C(C(=O)O)N

Specifications

Melting Point 182.0°C to 192.0°C
Color White
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Quantity 25 g
Beilstein 04,II,931
Merck Index 15,4511
Specific Rotation Condition − 16.50 (20.00°C c=2,H2O)
Specific Rotation − 16.50
Solubility Information Solubility in water: very soluble. Other solubilities: freely soluble in dilute alcohol and dimethyl-, formamide, almost insoluble in ethanol
Formula Weight 307.32
Percent Purity 98%
Grade Analysis
Physical Form Crystalline Powder
Chemical Name or Material Glutathione, For analysis, Reduced
Show More Show Less
EINECSNumber 200-725-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.